Learn More
2-Aminobenzimidazole, 97+%, Thermo Scientific Chemicals
Catalog Number p-7043686
Click to view available options
Quantity:
5 g
25 g
100 g
934-32-7
C7H7N3
133.15
MFCD00005596
JWYUFVNJZUSCSM-UHFFFAOYSA-N
2-aminobenzimidazole, 1h-benzo d imidazol-2-amine, 2-iminobenzimidazoline, benzimidazole, 2-amino, benzimidazol-2-ylamine, 2-amino benzimidazole, caswell no. 033aa, 1h-1,3-benzodiazol-2-amine, usaf ek-4037, 2-amino-1h-benzimidazole
13624
CHEBI:27822
1H-benzimidazol-2-amine
NC1=NC2=CC=CC=C2N1
934-32-7
C7H7N3
133.15
MFCD00005596
JWYUFVNJZUSCSM-UHFFFAOYSA-N
2-aminobenzimidazole, 1h-benzo d imidazol-2-amine, 2-iminobenzimidazoline, benzimidazole, 2-amino, benzimidazol-2-ylamine, 2-amino benzimidazole, caswell no. 033aa, 1h-1,3-benzodiazol-2-amine, usaf ek-4037, 2-amino-1h-benzimidazole
13624
CHEBI:27822
1H-benzimidazol-2-amine
NC1=NC2=CC=CC=C2N1
2-Aminobenzimidazole is used as receptors in chemosensor analysis. It was used in the hydrolysis of a choline carbonate. It was also used in the synthesis of imidazo[1,2-a]benzimidazoles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Aminobenzimidazole is used as receptors in chemosensor analysis. It was used in the hydrolysis of a choline carbonate. It was also used in the synthesis of imidazo[1,2-a]benzimidazoles.
Solubility
Soluble in water (Slightly).
Notes
Protect from direct light. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY
Melting Point 228°C to 232°C
Quantity 5 g
Beilstein 116525
Solubility Information Soluble in water (Slightly).
Formula Weight 133.15
Percent Purity ≥97%
Chemical Name or Material 2-Aminobenzimidazole