Learn More
2-Aminobenzoxazole, TRC
Specifications
Specifications
| Chemical Name or Material | 1,3-Benzoxazol-2-amine |
| CAS | 4570-41-6 |
| Molecular Formula | C7 H6 N2 O |
| Quantity | 1 g |
| Synonym | 2-Benzoxazolamine, 1,3-Benzoxazol-2-amine |
| InChI Formula | InChI=1S/C7H6N2O/c8-7-9-5-3-1-2-4-6(5)10-7/h1-4H,(H2,8,9) |
| SMILES | Nc1oc2ccccc2n1 |
| IUPAC Name | 1,3-benzoxazol-2-amine |
| Molecular Weight (g/mol) | 134.14 |
| Formula Weight | 134.048 |