Läs mer
2-Aminobenzylcyanide, Hydrochloride, TRC
Produktkod. 30322899
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30322899 500 mg
1 options
Produktkod. 30322899 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA596925500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Aminobenzylcyanide, Hydrochloride
CAS 40943-70-2
Molekylformel C8 H8 N2 . Cl H
Kvantitet 500 mg
Synonym Benzeneacetonitrile, 2-amino-, hydrochloride (1:1), Benzeneacetonitrile, 2-amino-, monohydrochloride (9CI), o-Aminophenylacetonitrile hydrochloride, 2-Aminobenzeneacetonitrile hydrochloride, (2-Aminophenyl)acetonitrile hydrochloride, 2-(Cyanomethyl)aniline hydrochloride, 2-Aminobenzyl cyanide hydrochloride, o-Aminobenzyl cyanide hydrochloride
InChI formel InChI=1S/C8H8N2.ClH/c9-6-5-7-3-1-2-4-8(7)10;/h1-4H,5,10H2;1H
LEDER Cl.Nc1ccccc1CC#N
IUPAC-namn 2-(2-aminophenyl)acetonitrile;hydrochloride
Molekylvikt (g/mol) 168.62
Formel vikt 168.0454