Läs mer
2-Aminoethyl 2,3-dihydroxypropyl Hydrogen Phosphate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | L-Alpha-Glycerophosphorylethanolamine |
| CAS | 33049-08-0 |
| Anteckningar om renhetsgrad | HPLC |
| Molekylformel | C5 H14 N O6 P |
| Kvantitet | 250 mg |
| Synonym | Phosphoric acid, mono(2-aminoethyl) mono[(2R)-2,3-dihydroxypropyl] ester, Phosphoric acid, mono(2-aminoethyl) mono(2,3-dihydroxypropyl) ester, (R)-, Phosphoric acid, mono(2-aminoethyl) mono(2,3-dihydroxypropyl) ester, D- (8CI), L-α-Glycerophosphorylethanolamine, Phosphoric acid mono(2-aminoethyl) mono[(2R)-2,3-dihydroxypropyl] ester, 2-Aminoethyl [(2R)-2,3-dihydroxypropyl] hydrogen phosphate, 2-Aminoethyl 2,3-dihydroxypropyl Hydrogen Phosphate |
| InChI formel | InChI=1S/C5H14NO6P/c6-1-2-11-13(9,10)12-4-5(8)3-7/h5,7-8H,1-4,6H2,(H,9,10)/t5-/m1/s1 |
| LEDER | NCCOP(=O)(O)OC[C@H](O)CO |
| IUPAC-namn | 2-aminoethyl [(2R)-2,3-dihydroxypropyl] hydrogen phosphate |
| Molekylvikt (g/mol) | 215.142 |
| Visa mer |