Learn More
2-Aminoethyl methacrylate hydrochloride, 90%, stabilized, Thermo Scientific Chemicals
Catalog Number p-2689922
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2420-94-2
C6H11NO2·HCl
165.62
MFCD00078260
XSHISXQEKIKSGC-UHFFFAOYSA-N
2-aminoethyl methacrylate hydrochloride, 2-aminoethylmethacrylate hydrochloride, 2-aminoethyl 2-methylacrylate hydrochloride, methacrylic acid, 2-aminoethyl ester, hydrochloride, polymers, 2-aminoethyl 2-methylprop-2-enoate hydrochloride, 2-propenoic acid, 2-methyl-, 2-aminoethyl ester, hydrochloride 1:1, 2-propenoic acid, 2-methyl-, 2-aminoethyl ester, hydrochloride, aminoethylmethacrylate, acmc-209wgm, timtec-bb sbb003905
75495
2-aminoethyl 2-methylprop-2-enoate;hydrochloride
CC(=C)C(=O)OCCN.Cl
2420-94-2
C6H11NO2·HCl
165.62
MFCD00078260
XSHISXQEKIKSGC-UHFFFAOYSA-N
2-aminoethyl methacrylate hydrochloride, 2-aminoethylmethacrylate hydrochloride, 2-aminoethyl 2-methylacrylate hydrochloride, methacrylic acid, 2-aminoethyl ester, hydrochloride, polymers, 2-aminoethyl 2-methylprop-2-enoate hydrochloride, 2-propenoic acid, 2-methyl-, 2-aminoethyl ester, hydrochloride 1:1, 2-propenoic acid, 2-methyl-, 2-aminoethyl ester, hydrochloride, aminoethylmethacrylate, acmc-209wgm, timtec-bb sbb003905
75495
2-aminoethyl 2-methylprop-2-enoate;hydrochloride
CC(=C)C(=O)OCCN.Cl
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
CAS Min % 88.0
CAS Max % 100.0
Melting Point 116.0°C to 118.0°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 88% min. (Argentometry)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: soluble
Stabilizer 500 to 1000ppm phenothiazine
Formula Weight 165.62
Percent Purity 90%
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminoethyl methacrylate hydrochloride, 90%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Aminoethyl methacrylate hydrochloride, stabilized
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.