Läs mer
(±)-2-Aminohexane-1,1,1,2,3,3-d6, TRC
Produktkod. 30328579
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30328579 10 mg
30324809 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30328579 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA21129610MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Hexanamine-1,1,1,2,3,3-D6
CAS 1219802-28-4
Molekylformel C6 D6 H9 N
Kvantitet 10 mg
Synonym 1-(Methyl-d3)pentyl-1,2,2-d3-amine, 2-Aminohexane-1,1,1,2,3,3-d6, 2-Hexanamine-1,1,1,2,3,3-D6, (±)-2-Aminohexane-1,1,1,2,3,3-d6
InChI formel InChI=1S/C6H15N/c1-3-4-5-6(2)7/h6H,3-5,7H2,1-2H3/i2D3,5D2,6D
LEDER [2H]C([2H])([2H])C([2H])(N)C([2H])([2H])CCC
IUPAC-namn 1,1,1,2,3,3-hexadeuteriohexan-2-amine
Molekylvikt (g/mol) 107.227
Formel vikt 107.1581