missing translation for 'onlineSavingsMsg'
Learn More

2-Aminopyridine, 99%, Thermo Scientific Chemicals

Catalog Number p-7082099
Click to view available options
Quantity:
25 g
250 g
1000 g
This item is not returnable. View return policy
504-29-0
C5H6N2
94.117
MFCD00006312
ICSNLGPSRYBMBD-UHFFFAOYSA-N
2-aminopyridine, 2-pyridinamine, 2-pyridylamine, aminopyridine, pyridinamine, o-aminopyridine, 2-aminopyridin, 2-aminopryidine, amino-2 pyridine, pyridine, 2-amino
10439
pyridin-2-amine
C1=CC=NC(=C1)N
This item is not returnable. View return policy
504-29-0
C5H6N2
94.117
MFCD00006312
ICSNLGPSRYBMBD-UHFFFAOYSA-N
2-aminopyridine, 2-pyridinamine, 2-pyridylamine, aminopyridine, pyridinamine, o-aminopyridine, 2-aminopyridin, 2-aminopryidine, amino-2 pyridine, pyridine, 2-amino
10439
pyridin-2-amine
C1=CC=NC(=C1)N

A compound has been shown to reversibly block voltage-dependent potassium channels. Also a common impurity from the synthesis of compounds found in hair dye. Used as a water miscible solvent and as an intermediate in manufacturing pharmaceuticals like diuretics. It is used as a research tool, in characterizing subtypes of the potassium channel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A compound has been shown to reversibly block voltage-dependent potassium channels. Also a common impurity from the synthesis of compounds found in hair dye. Used as a water miscible solvent and as an intermediate in manufacturing pharmaceuticals like diuretics. It is used as a research tool, in characterizing subtypes of the potassium channel.

Solubility
Slightly soluble in water(8g/100 mL at 19°C)

Notes
Hygroscopic. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acids.
TRUSTED_SUSTAINABILITY
Melting Point 56°C to 60°C
Boiling Point 209°C to 211°C
Flash Point 92°C (197°F)
Quantity 25 g
UN Number UN2671
Beilstein 105785
Sensitivity Hygroscopic
Merck Index 14,473
Solubility Information Slightly soluble in water(8g/100 mL at 19°C)
Formula Weight 94.12
Percent Purity 99%
Chemical Name or Material 2-Aminopyridine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'