Läs mer
2-(Aminosulfonyl)benzoic Acid, TRC
Produktkod. 30310354
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30310354 25 mg
1 options
Produktkod. 30310354 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA62936025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Carboxybenzenesulfonamide
CAS 632-24-6
Molekylformel C7 H7 N O4 S
Kvantitet 25 mg
Synonym 2-(Aminosulfonyl)benzoic acid (ACI), Benzoic acid, o-sulfamoyl- (6CI, 8CI), 2-Carboxybenzenesulfonamide, 2-Sulfamoylbenzoic acid, NSC 22975, o-Sulfamidobenzoic acid, o-Sulfamoylbenzoic acid, o-Sulfonamidobenzoic acid, IN-D5119
InChI formel InChI=1S/C7H7NO4S/c8-13(11,12)6-4-2-1-3-5(6)7(9)10/h1-4H,(H,9,10)(H2,8,11,12)
LEDER NS(=O)(=O)c1ccccc1C(=O)O
IUPAC-namn 2-sulfamoylbenzoic acid
Molekylvikt (g/mol) 201.2
Formel vikt 201.01