Läs mer
2-Arachidonoyl-sn-glycero-3-phosphocholine (>90%), TRC
Produktkod. 30137043
Change view
Kvantitet:
2.5 mg
10 mg
25 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30137043 25 mg -
30132901 2.5 mg 2.5mg
30011371 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137043 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA76509025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 67341-29-1
Molekylformel C28H50NO7P
Molekylvikt (g/mol) 543.68
Synonym (7R,13Z,16Z,19Z,22Z)-4-Hydroxy-7-(hydroxymethyl)-N,N,N-trimethyl-9-oxo-3,5,8-trioxa-4-phosphaoctacosa-13,16,19,22-tetraen-1-aminium 4-Oxide Inner Salt, [R-(all-Z)]-4-Hydroxy-7-(hydroxymethyl)-N,N,N-trimethyl-9-oxo-3,5,8-trioxa-4-phosphaoctacosa-13,16,19,22-tetraen-1-aminium 4-Oxide Inner Salt
IUPAC-namn (R)-3-hydroxy-2-(((5Z,8Z,11Z,14Z)-icosa-5,8,11,14-tetraenoyl)oxy)propyl (2-(trimethylammonio)ethyl) phosphate
LEDER O=C(CCC/C=C\C/C=C\C/C=C\C/C=C\CCCCC)O[C@@H](COP([O-])(OCC[N+](C)(C)C)=O)CO
Specifikationer
Kemiskt namn eller material 2-Arachidonoyl-sn-glycero-3-phosphocholine
Analyt- eller komponentnamn 2-Arachidonoyl-sn-glycero-3-phosphocholine
Frakt skick Dry ice
InChI formel InChI=1S/C28H50NO7P/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-28(31)36-27(25-30)26-35-37(32,33)34-24-23-29(2,3)4/h9-10,12-13,15-16,18-19,27,30H,5-8,11,14,17,20-26H2,1-4H3/b10-9-,13-12-,16-15-,19-18-/t27-/m1/s1
Lösningstyp Neat
Industry Type Pharmaceutical
Kvantitet 25 mg
Formel vikt 543.33