missing translation for 'onlineSavingsMsg'
Läs mer

2-Azabicyclo[2.2.2]octan-3-one, TRC

Produktkod. 30340053
Change view
Click to view available options
Kvantitet:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30340053 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30340053 Supplier Toronto Research Chemicals Supplier No. TRCA96579850MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 2-Azabicyclo[2.2.2]octan-3-one
CAS 3306-69-2
Molekylformel C7H11NO
Kvantitet 50 mg
Synonym 3-Isoquinuclidinone, 3-Isoquinuclidone, 4-Aminocyclohexanecarboxylic acid lactam, Isonortropinone, NSC 85851
InChI formel InChI=1S/C7H11NO/c9-7-5-1-3-6(8-7)4-2-5/h5-6H,1-4H2,(H,8,9)
LEDER O=C1NC2CCC1CC2
IUPAC-namn 2-azabicyclo[2.2.2]octan-3-one
Molekylvikt (g/mol) 125.17
Formel vikt 125.0841
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.