Läs mer
2-Azido-2-deoxy-D-glucose, TRC
Produktkod. 17943345
Change view
Kvantitet:
100 mg
500 mg
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17943345 100 mg
17900395 500 mg
17910735 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17943345 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA824100100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 56883-39-7
Molekylformel C6 H11 N3 O5
Molekylvikt (g/mol) 205.17
IUPAC-namn (3R,4R,5S,6R)-3-azido-6-(hydroxymethyl)oxane-2,4,5-triol
LEDER OC[C@H]1OC(O)[C@H](N=[N+]=[N-])[C@@H](O)[C@@H]1O
Specifikationer
Kemiskt namn eller material 2-Azido-2-deoxy-D-glucose
Kvantitet 100 mg
InChI formel InChI=1S/C6H11N3O5/c7-9-8-3-5(12)4(11)2(1-10)14-6(3)13/h2-6,10-13H,1H2/t2-,3-,4-,5-,6?/m1/s1
Formel vikt 205.0699