Läs mer
2'-Benzoyl-2,4'-dichloroacetanilide, Mikromol™
Produktkod. 30051895
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30051895 25 mg
17973896 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30051895 Leverantör Mikromol Leverantörsnummer MM0023.050025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2'-Benzoyl-2,4'-dichloroacetanilide
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2'-Benzoyl-2,4'-dichloroacetanilide
CAS 4016-85-7
Analyt- eller komponentnamn 2'-Benzoyl-2,4'-dichloroacetanilide
Frakt skick Room Temperature
Molekylformel C15 H11 Cl2 N O2
Certifikat och efterlevnad ISO 17025
Synonym 2'-Benzoyl-2,4'-dichloroacetanilide
InChI formel InChI=1S/C15H11Cl2NO2/c16-9-14(19)18-13-7-6-11(17)8-12(13)15(20)10-4-2-1-3-5-10/h1-8H,9H2,(H,18,19)
LEDER ClCC(=O)Nc1ccc(Cl)cc1C(=O)c2ccccc2
IUPAC-namn N-(2-benzoyl-4-chlorophenyl)-2-chloroacetamide
Molekylvikt (g/mol) 308.16
Kvantitet 25 mg
Formel vikt 307.0167 g/mol