Läs mer
2-Benzoyl-5-ethylthiophene, TRC
Produktkod. 30330774
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30330774 1 g
1 options
Produktkod. 30330774 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB4419101G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Decarboxylated Tiaprofenic Acid
CAS 6933-26-2
Molekylformel C13 H12 O S
Kvantitet 1 g
Synonym Methanone, (5-ethyl-2-thienyl)phenyl-, Ketone, 5-ethyl-2-thienyl phenyl (7CI,8CI), (5-Ethyl-2-thienyl)phenylmethanone, (5-Ethylthiophen-2-yl)(phenyl)methanone, 2-Benzoyl-5-ethylthiophene, Decarboxylated tiaprofenic acid, Tiaprofenic Acid Imp. A (EP)
InChI formel InChI=1S/C13H12OS/c1-2-11-8-9-12(15-11)13(14)10-6-4-3-5-7-10/h3-9H,2H2,1H3
LEDER CCc1ccc(s1)C(=O)c2ccccc2
IUPAC-namn (5-ethylthiophen-2-yl)-phenylmethanone
Molekylvikt (g/mol) 216.299
Formel vikt 216.061