Läs mer
2-Benzyl-2-(dimethylamino)-4'-morpholinobutyrophenone, TRC
Produktkod. 30347297
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30347297 250 mg
1 options
Produktkod. 30347297 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB535388250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-(Dimethylamino)-1-[4-(4-morpholinyl)phenyl]-2-(phenylmethyl)-1-butanone
CAS 119313-12-1
Molekylformel C23 H30 N2 O2
Kvantitet 250 mg
Synonym 2-(Dimethylamino)-1-[4-(4-morpholinyl)phenyl]-2-(phenylmethyl)-1-butanone, 2-(Dimethylamino)-1-(4-morpholinophenyl)-2-benzyl-1-butanone, 2-Benzyl-2-(dimethylamino)-1-(4-morpholinophenyl)-1-butanone, 2-Benzyl-2-(dimethylamino)-1-(4-morpholinophenyl)butanone-1, 2-Benzyl-2-(dimethylamino)-1-[4-(4-morpholinyl)phenyl]-1-butanone, 2-Benzyl-2-(dimethylamino)-4'-mopholinobutyrophenone, 2-Benzyl-2-(dimethylamino)-4'-morpholinobutyrophenone, 2-Benzyl-2-N,N-dimethylamino-1-(4-morpholinophenyl)-1-butanone, 2-Benzyl-2-dimethylamino-1-(4-morpholinophehyl)-butane-1-one, 2-Benzyl-2-dimethylamino-1-(4-morpholinophenyl)butanone, 2-Dimethylamino-2-benzyl-1-[4-(4-morpholino)phenyl]-1-butanone, CGI 369, Ciba 369, Genocure BDMM, I 369, IC 369, IGR 369, IHT-PI 910, Irg 369, Irgacure 369, Irgacure 369E, JRcure 369, Lancure 369, Luna 800, Omnired 369, PI 369, Photoinitiator 369, UV 369, α-Benzyl-α-(dimethylamino)-4-morpholinobutyrophenone
InChI formel InChI=1S/C23H30N2O2/c1-4-23(24(2)3,18-19-8-6-5-7-9-19)22(26)20-10-12-21(13-11-20)25-14-16-27-17-15-25/h5-13H,4,14-18H2,1-3H3
LEDER CCC(Cc1ccccc1)(N(C)C)C(=O)c2ccc(cc2)N3CCOCC3
IUPAC-namn 2-benzyl-2-(dimethylamino)-1-(4-morpholin-4-ylphenyl)butan-1-one
Molekylvikt (g/mol) 366.5
Formel vikt 366.2307