Läs mer
2-Benzylamino-2-phenylbutanol, TRC
Produktkod. 30333645
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30333645 10 mg
1 options
Produktkod. 30333645 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB22599010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Benzylamino-2-phenylbutanol
CAS 1178412-63-9
Molekylformel C17H21NO
Kvantitet 10 mg
Synonym N-Benzyl 1-Phenyl-1-hydroxymetbl-1-propanamine, beta-Ethyl-beta-[(phenylmethyl)amino]benzeneethanol
InChI formel InChI=1S/C17H21NO/c1-2-17(14-19,16-11-7-4-8-12-16)18-13-15-9-5-3-6-10-15/h3-12,18-19H,2,13-14H2,1H3
LEDER CCC(CO)(NCc1ccccc1)c2ccccc2
IUPAC-namn 2-(benzylamino)-2-phenylbutan-1-ol
Molekylvikt (g/mol) 255.35
Formel vikt 255.1623