Learn More
2-Benzylbenzimidazole Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Benzylbenzimidazole Hydrochloride |
| CAS | 1212-48-2 |
| Molecular Formula | C14H12N2 •HCl |
| Quantity | 1 g |
| Synonym | Dibasole, Dibazol Hydrochloride, Dibazole Hydrochloride, Bendazol Hydrochloride, 2-(Phenylmethyl)-1H-benzimidazole Monohydrochloride, Benzimidazole, 2-benzyl-, Monohydrochloride (8CI), 1H-Benzimidazole, 2-(phenylmethyl)-, Monohydrochloride (9CI) |
| InChI Formula | InChI=1S/C14H12N2.ClH/c1-2-6-11(7-3-1)10-14-15-12-8-4-5-9-13(12)16-14;/h1-9H,10H2,(H,15,16);1H |
| SMILES | C1=CC=C(C=C1)CC2=NC3=CC=CC=C3N2.Cl |
| IUPAC Name | 2-benzyl-1H-benzimidazole;hydrochloride |
| Molecular Weight (g/mol) | 208.263646 |