Läs mer
2-(Benzylethyl-amino)-1-(3-hydroxyphenyl)ethanone Hydrochloride (Benzyletilefrone Hydrochloride), Mikromol™
Produktkod. 30049901
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30049901 25 mg
30049316 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30049901 Leverantör Mikromol Leverantörsnummer MM0035.080025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-(Benzylethyl-amino)-1-(3-hydroxyphenyl)ethanone Hydrochloride (Benzyletilefrone Hydrochloride)
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-(Benzylethyl-amino)-1-(3-hydroxyphenyl)ethanone Hydrochloride (Benzyletilefrone Hydrochloride)
CAS 55845-90-4
Analyt- eller komponentnamn 2-(Benzylethyl-amino)-1-(3-hydroxyphenyl)ethanone Hydrochloride (Benzyletilefrone Hydrochloride)
Frakt skick Room Temperature
Molekylformel C17 H19 N O2 . Cl H
Certifikat och efterlevnad ISO 9001
Synonym Etilefrine Hydrochloride Imp. D (EP), Etilefrine Imp. D (EP), 2-(Benzylethyl-amino)-1-(3-hydroxyphenyl)ethanone Hydrochloride, Etilefrine Hydrochloride Imp. D (EP) as Hydrochloride, Benzyletilefrone Hydrochloride, Etilefrine Hydrochloride Impurity D as Hydrochloride, Etilefrine Impurity D as Hydrochloride
InChI formel InChI=1S/C17H19NO2.ClH/c1-2-18(12-14-7-4-3-5-8-14)13-17(20)15-9-6-10-16(19)11-15;/h3-11,19H,2,12-13H2,1H3;1H
LEDER Cl.CCN(CC(=O)c1cccc(O)c1)Cc2ccccc2
IUPAC-namn 2-[benzyl(ethyl)amino]-1-(3-hydroxyphenyl)ethanone;hydrochloride
Molekylvikt (g/mol) 305.8
Kvantitet 25 mg
Formel vikt 305.1183 g/mol