Läs mer
2-(Benzyloxy)acetamide, TRC
Produktkod. 30337811
Change view
Kvantitet:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30337811 250 mg
30330634 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30337811 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB288563250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-(Benzyloxy)acetamide
CAS 5774-77-6
Molekylformel C9H11NO2
Kvantitet 250 mg
InChI formel InChI=1S/C9H11NO2/c10-9(11)7-12-6-8-4-2-1-3-5-8/h1-5H,6-7H2,(H2,10,11)
LEDER NC(=O)COCc1ccccc1
IUPAC-namn 2-phenylmethoxyacetamide
Molekylvikt (g/mol) 165.19
Formel vikt 165.079