Läs mer
2-Biphenylyl diphenyl phosphate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | 2-Biphenylyl diphenyl phosphate |
| CAS | 132-29-6 |
| Anteckningar om renhetsgrad | HPLC |
| Molekylformel | C24H19O4P |
| Kvantitet | 500 mg |
| Synonym | Diphenyl mono-o-xenyl phosphate, Diphenyl o-biphenylyl phosphate, Dow 5, Dow Plasticizer 5, NSC 2889, o-Phenylphenyl bisphenyl phosphate, o-Phenylphenyl diphenyl phosphate, o-Xelenyl diphenyl phosphate |
| InChI formel | InChI=1S/C24H19O4P/c25-29(26-21-14-6-2-7-15-21,27-22-16-8-3-9-17-22)28-24-19-11-10-18-23(24)20-12-4-1-5-13-20/h1-19H |
| LEDER | O=P(Oc1ccccc1)(Oc2ccccc2)Oc3ccccc3c4ccccc4 |
| IUPAC-namn | diphenyl (2-phenylphenyl) phosphate |
| Molekylvikt (g/mol) | 402.38 |
| Visa mer |