Läs mer
2-Biphenylyl Sulfate Potassium Salt, TRC
Produktkod. 30346762
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30346762 50 mg
1 options
Produktkod. 30346762 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB39789550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Biphenylyl Sulfate Potassium Salt
CAS 854243-47-3
Molekylformel C12H10KO4S
Kvantitet 50 mg
Synonym [1,1'-Biphenyl]-2-ol, 2-(hydrogen Sulfate) Potassium Salt
LEDER [K].O=S(=O)(O)OC=1C=CC=CC1C=2C=CC=CC2
Molekylvikt (g/mol) 289.37