Learn More
2-Biphenylylamine, Sigma-Aldrich™
Product Code. 15850781
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15850781 50 g 50g
15840781 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15850781 Supplier Sigma Aldrich Supplier No. 8201550050
 Restricted Item
Beräknad leverans: 19-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 90-41-5
Grade Research
Molecular Formula C6H5C6H4-2-(NH2)
Molecular Weight (g/mol) 169.22
Specifications
Melting Point 46°C to 48°C
Boiling Point 298°C to 300°C
MDL Number MFCD00007701
Quantity 50 g
Synonym 2-Biphenylylamine, 2-Aminodiphenyl, 2-Phenylaniline, 2-Biphenylylamine