missing translation for 'onlineSavingsMsg'
Learn More

2-Boc-2-azabicyclo[2.2.1]hept-5-ene, 98%, Thermo Scientific Chemicals

Catalog Number 15460529
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
188345-71-3
C11H17NO2
195.262
MFCD22416354
ZATXHTCUZAWODK-UHFFFAOYSA-N
15837921
tert-butyl 5-azabicyclo[2.2.1]hept-2-ene-5-carboxylate
CC(C)(C)OC(=O)N1CC2CC1C=C2
This item is not returnable. View return policy
188345-71-3
C11H17NO2
195.262
MFCD22416354
ZATXHTCUZAWODK-UHFFFAOYSA-N
15837921
tert-butyl 5-azabicyclo[2.2.1]hept-2-ene-5-carboxylate
CC(C)(C)OC(=O)N1CC2CC1C=C2

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Yellow
Quantity 250 mg
Synonym 2-boc-2-azabicyclo 2.2.1 hept-5-ene, tert-butyl 2-aza-bicyclo 2.2.1 hept-5-ene-2-carboxylate, tert-butyl 2-azabicyclo 2.2.1 hept-5-ene-2-carboxylate, 2-tert-butoxycarbonyl-2-azabicyclo 2.2.1 hept-5-ene, tert-butyl2-azabicyclo 2.2.1 hept-5-ene-2-carboxylate, ksc497c1n, tert-butyl 5-azabicyclo 2.2.1 hept-2-ene-5-carboxylate, 2-aza-bicyclo 2.2.1 hept-5-ene-2-carboxylic acid tert-butyl ester, 2-tert-butyloxycarbonyl-2-azabicyclo 2.2.1 hepta-5-ene, 2-azabicyclo 2.2.1 hept-5-ene-2-carboxylic acid tert-butyl ester
Formula Weight 195.26
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Boc-2-azabicyclo[2.2.1]hept-5-ene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'