missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-(1-methyl-1H-benzimidazol-5-yl)ethanone hydrobromide, 97%, Thermo Scientific™

Catalog Number p-3586654
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
944450-78-6
C10H10Br2N2O
334.011
MFCD09817461
LRWGKOQDEBXXPS-UHFFFAOYSA-N
2-bromo-1-1-methyl-1h-benzimidazol-5-yl ethanone hydrobromide, 2-bromo-1-1-methyl-1h-benzo d imidazol-5-yl ethanone hydrobromide, 2-bromo-1-1-methyl-1h-benzimidazol-5-yl ethan-1-one hydrobromide, 2-bromo-1-1-methyl-1,3-benzodiazol-5-yl ethanone hydrobromide, c10h9brn2o.hbr, 2-bromo-1-1-methyl-1h-benzoimidazol-5-yl-ethanone.hbr, 2-bromo-1-1-methyl-1h-benzo d imidazol-5-yl ethan-1-one hydrobromide, 2-bromo-1-1-methyl-1h-benzimidazol-5-yl ethan-1-one-hydrogen bromide 1/1
24229460
2-bromo-1-(1-methylbenzimidazol-5-yl)ethanone;hydrobromide
CN1C=NC2=C1C=CC(=C2)C(=O)CBr.Br
This item is not returnable. View return policy
944450-78-6
C10H10Br2N2O
334.011
MFCD09817461
LRWGKOQDEBXXPS-UHFFFAOYSA-N
2-bromo-1-1-methyl-1h-benzimidazol-5-yl ethanone hydrobromide, 2-bromo-1-1-methyl-1h-benzo d imidazol-5-yl ethanone hydrobromide, 2-bromo-1-1-methyl-1h-benzimidazol-5-yl ethan-1-one hydrobromide, 2-bromo-1-1-methyl-1,3-benzodiazol-5-yl ethanone hydrobromide, c10h9brn2o.hbr, 2-bromo-1-1-methyl-1h-benzoimidazol-5-yl-ethanone.hbr, 2-bromo-1-1-methyl-1h-benzo d imidazol-5-yl ethan-1-one hydrobromide, 2-bromo-1-1-methyl-1h-benzimidazol-5-yl ethan-1-one-hydrogen bromide 1/1
24229460
2-bromo-1-(1-methylbenzimidazol-5-yl)ethanone;hydrobromide
CN1C=NC2=C1C=CC(=C2)C(=O)CBr.Br
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 334.01
Percent Purity 97%
Chemical Name or Material 2-Bromo-1-(1-methyl-1H-benzimidazol-5-yl)ethanone hydrobromide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'