Läs mer
2-Bromo-1-(2-pyrazinyl) Ethanone Hydrobromide, TRC
Produktkod. 30345586
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30345586 100 mg
1 options
Produktkod. 30345586 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB686900100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Bromo-1-(2-pyrazinyl) Ethanone Hydrobromide
CAS 126353-32-0
Molekylformel C6H6Br2N2O
Kvantitet 100 mg
Synonym 2-Bromo-1-pyrazin-2-ylethanone, 2-Bromo-1-pyrazinylethanone Hydrobromide, 2-(2-Bromoacetyl)pyrazine, 2-(Bromoacetyl)pyrazine Hydrobromide
InChI formel InChI=1S/C6H5BrN2O.BrH/c7-3-6(10)5-4-8-1-2-9-5;/h1-2,4H,3H2;1H
LEDER Br.BrCC(=O)c1cnccn1
IUPAC-namn 2-bromo-1-pyrazin-2-ylethanone;hydrobromide
Molekylvikt (g/mol) 281.93
Formel vikt 279.8847