missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1,3,4-trifluorobenzene, 97%, Thermo Scientific™

Catalog Number 11469646
Click to view available options
Quantity:
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS 176793-04-7
Flash Point 62°C (143°F)
Molecular Formula C6H2BrF3
Refractive Index 1.4895
MDL Number MFCD00236214
Quantity 10 g
Synonym 1-bromo-2,3,6-trifluorobenzene, 2,3,6-trifluorobromobenzene, benzene, 2-bromo-1,3,4-trifluoro, 3-bromo-1,2,4-trifluorobenzene, 2-bromo-1,3,4-trifluoro-benzene, pubchem8527, 1,3,4-tifluorobenzene, acmc-209ec0, 2,3,6-trifluoro-bromobenzene, 2,3,6-trifluorophenyl bromide
InChI Key CNAXDCSHYHIOGW-UHFFFAOYSA-N
SMILES FC1=CC=C(F)C(Br)=C1F
IUPAC Name 2-bromo-1,3,4-trifluorobenzene
Molecular Weight (g/mol) 210.98
PubChem CID 2736401
Formula Weight 210.98
Percent Purity 97%
Chemical Name or Material 2-Bromo-1,3,4-trifluorobenzene
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'