missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-(3-thienyl)-1-ethanone, 97%, Thermo Scientific™

Catalog Number p-4510104
Click to view available options
Quantity:
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
1468-82-2
C6H5BrOS
205.07
MFCD02677695
TXEJYUFJFSPCHH-UHFFFAOYSA-N
2776379
BrCC(=O)C1=CSC=C1
This item is not returnable. View return policy
1468-82-2
C6H5BrOS
205.07
MFCD02677695
TXEJYUFJFSPCHH-UHFFFAOYSA-N
2776379
BrCC(=O)C1=CSC=C1
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Synonym 2-bromo-1-3-thienyl-1-ethanone, 2-bromo-1-thiophen-3-yl ethanone, 3-bromoacetylthiophene, 3-bromoacetyl thiophene, 2-bromo-1-thiophen-3-yl ethan-1-one, 2-bromo-1-3-thienyl ethanone, ethanone,2-bromo-1-3-thienyl, ethanone, 2-bromo-1-3-thienyl, 2-bromo-1-thien-3-yl-1-ethanone
IUPAC Name 2-bromo-1-thiophen-3-ylethanone
Formula Weight 205.07
Percent Purity 97%
Chemical Name or Material 2-Bromo-1-(3-thienyl)-1-ethanone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'