Learn More
2-Bromo-1-(4-hydroxyphenyl)ethanone, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 2491-38-5 |
| Molecular Formula | C8H7BrO2 |
| MDL Number | MFCD00072424 |
| Quantity | 25 g |
| SMILES | OC1=CC=C(C=C1)C(=O)CBr |
| Molecular Weight (g/mol) | 215.04 |
| Percent Purity | 98% |
| Chemical Name or Material | 2-Bromo-1-(4-hydroxyphenyl)ethanone |