Läs mer
2-brom-1-(4-morfolinofenyl)-1-etanon, 97 %, Thermo Scientific™
Produktkod. 10394562
Change view
Kvantitet:
250 mg
1 g
5 g
Förpackningsstorlek:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
10394562 250 mg 250mg
10792473 1 g 1g
10275244 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 10394562 Leverantör Thermo Scientific Maybridge Leverantörsnummer CC17405CB
Kemiska identifierare
CAS 210832-85-2
Molekylformel C12H14BrNO2
Molekylvikt (g/mol) 284.15
MDL-nummer MFCD03783555
InChI-nyckel OUGMZFJPRSTGMJ-UHFFFAOYSA-N
Synonym 2-bromo-1-4-morpholinophenyl-1-ethanone, 2-bromo-1-4-morpholinophenyl ethanone, 2-bromo-1-4-morpholin-4-ylphenyl ethanone, 2-bromo-1-4-4-morpholinyl phenyl ethanone, 2-bromo-1-4-morpholin-4-yl-phenyl ethanone, 2-bromo-1-4-morpholin-4-yl phenyl ethanone, 2-bromo-1-4-morpholin-4-ylphenyl ethan-1-one, pubchem23338, acmc-1cqxm, alpha-bromo-4'-morpholinoacetophenone
PubChem CID 2795357
LEDER BrCC(=O)C1=CC=C(C=C1)N1CCOCC1
Specifikationer
Kemiskt namn eller material 2-Bromo-1-(4-morpholinophenyl)-1-ethanone
Analysprocentintervall 95%
Kvantitet 250 mg
IUPAC-namn 2-brom-l-(4-morfolin-4-ylfenyl)etanon
Formel vikt 284.15
Procent renhet 95%