missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-(4-phenoxyphenyl)ethanone 95+%, Thermo Scientific™

Catalog Number p-4510713
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
28179-33-1
C14H11BrO2
291.14
MFCD01419855
RAXTYMXDSNWNJS-UHFFFAOYSA-N
2-bromo-1-4-phenoxyphenyl ethanone, 2-bromo-1-4-phenoxyphenyl ethan-1-one, ethanone,2-bromo-1-4-phenoxyphenyl, 4-phenoxyphenacyl bromide, 2-bromo-4'-phenoxyacetophenone, 4'-phenoxy-2-bromoacetophenone, alpha-bromo-4'-phenoxyacetophenone, ethanone, 2-bromo-1-4-phenoxyphenyl
14797675
2-bromo-1-(4-phenoxyphenyl)ethanone
BrCC(=O)C1=CC=C(OC2=CC=CC=C2)C=C1
This item is not returnable. View return policy
28179-33-1
C14H11BrO2
291.14
MFCD01419855
RAXTYMXDSNWNJS-UHFFFAOYSA-N
2-bromo-1-4-phenoxyphenyl ethanone, 2-bromo-1-4-phenoxyphenyl ethan-1-one, ethanone,2-bromo-1-4-phenoxyphenyl, 4-phenoxyphenacyl bromide, 2-bromo-4'-phenoxyacetophenone, 4'-phenoxy-2-bromoacetophenone, alpha-bromo-4'-phenoxyacetophenone, ethanone, 2-bromo-1-4-phenoxyphenyl
14797675
2-bromo-1-(4-phenoxyphenyl)ethanone
BrCC(=O)C1=CC=C(OC2=CC=CC=C2)C=C1
Assay Percent Range 90%
Quantity 250 mg
Formula Weight 291.14
Percent Purity 90%
Chemical Name or Material 2-Bromo-1-(4-phenoxyphenyl)ethanone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'