missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-1-indanol, 99%, Thermo Scientific™
Click to view available options
Quantity:
10 g
50 g
250 g
Specifications
Melting Point | 130.0°C to 131.0°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (GC) |
Quantity | 10 g |
Formula Weight | 213.07 |
Percent Purity | 99% |
Chemical Name or Material | 2-Bromo-1-indanol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'