Läs mer
2-Bromo-3-benzyloxy-4-methoxybenzoic Acid Methyl Ester, TRC
Produktkod. 30347558
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30347558 50 mg
1 options
Produktkod. 30347558 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB68028050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Bromo-3-benzyloxy-4-methoxybenzoic Acid Methyl Ester
CAS 135586-19-5
Molekylformel C16H15BrO4
Kvantitet 50 mg
Synonym 2-Bromo-4-methoxy-3-(phenylmethoxy)-benzoic Acid Methyl Ester
InChI formel InChI=1S/C16H15BrO4/c1-19-13-9-8-12(16(18)20-2)14(17)15(13)21-10-11-6-4-3-5-7-11/h3-9H,10H2,1-2H3
LEDER COC(=O)c1ccc(OC)c(OCc2ccccc2)c1Br
IUPAC-namn methyl 2-bromo-4-methoxy-3-phenylmethoxybenzoate
Molekylvikt (g/mol) 351.19
Formel vikt 350.0154