Läs mer
2-Bromo-3-methoxy-6-nitropyridine, TRC
Produktkod. 30343237
Change view
Kvantitet:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30343237 100 mg
30344322 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30343237 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB684790100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Bromo-3-methoxy-6-nitropyridine
CAS 76066-07-4
Molekylformel C6 H5 Br N2 O3
Kvantitet 100 mg
InChI formel InChI=1S/C6H5BrN2O3/c1-12-4-2-3-5(9(10)11)8-6(4)7/h2-3H,1H3
LEDER COc1ccc(nc1Br)[N+](=O)[O-]
IUPAC-namn 2-bromo-3-methoxy-6-nitropyridine
Molekylvikt (g/mol) 233.02
Formel vikt 231.9484