missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-pyridinol, 99%, Thermo Scientific Chemicals

Catalog Number p-76417
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
6602-32-0
C5H4BrNO
174.00
MFCD00006220
YKHQFTANTNMYPP-UHFFFAOYSA-N
2-bromo-3-hydroxypyridine, 2-bromo-3-pyridinol, 3-pyridinol, 2-bromo, 2-bromo-pyridin-3-ol, 3-hydroxy-2-bromopyridine, unii-9dab85h359, 2-bromo-3-hydroxy pyridine, 2-bromo-3-hydroxy-pyridine, 2-bromo-3-hydroxypyridine 2-bromo-3-pyridinol, 2-bromo3-pyridinol
23070
2-bromopyridin-3-ol
OC1=CC=CN=C1Br
This item is not returnable. View return policy
6602-32-0
C5H4BrNO
174.00
MFCD00006220
YKHQFTANTNMYPP-UHFFFAOYSA-N
2-bromo-3-hydroxypyridine, 2-bromo-3-pyridinol, 3-pyridinol, 2-bromo, 2-bromo-pyridin-3-ol, 3-hydroxy-2-bromopyridine, unii-9dab85h359, 2-bromo-3-hydroxy pyridine, 2-bromo-3-hydroxy-pyridine, 2-bromo-3-hydroxypyridine 2-bromo-3-pyridinol, 2-bromo3-pyridinol
23070
2-bromopyridin-3-ol
OC1=CC=CN=C1Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 185°C to 188°C
Color Brown to Gray
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula Br(C5H3N)OH
Quantity 5 g
Formula Weight 174
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-3-pyridinol, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromo-3-pyridinol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'