missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Bromo-3-(trifluoromethyl)benzoic acid, 98%

Brand:  Alfa Aesar™ H63761.03

Additional Details : CAS Number : 177420-63-2 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15450619

  • 785.78 SEK / 1g


Estimated Shipment date
from Supplier 15-11-2018
Add to basket

Description

Description

Applications
2-Bromo-3-(trifluoromethyl)benzoic acid is used as a intermediate for pharmaceutical and chemical research.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

177420-63-2
269.017
FIMDBAHLQCCYJK-UHFFFAOYSA-N
15261012
C1=CC(=C(C(=C1)C(F)(F)F)Br)C(=O)O
C8H4BrF3O2
MFCD08532493
2-bromo-3-trifluoromethyl benzoic acid, benzoic acid, 2-bromo-3-trifluoromethyl, 2-bromo-3-carboxybenzotrifluoride, 2-bromo-3-trifluoromethylbenzoicacid
2-bromo-3-(trifluoromethyl)benzoic acid

Specifications

2-Bromo-3-(trifluoromethyl)benzoic acid
177420-63-2
MFCD08532493
FIMDBAHLQCCYJK-UHFFFAOYSA-N
2-bromo-3-(trifluoromethyl)benzoic acid
15261012
Solid
1g
C8H4BrF3O2
2-bromo-3-trifluoromethyl benzoic acid, benzoic acid, 2-bromo-3-trifluoromethyl, 2-bromo-3-carboxybenzotrifluoride, 2-bromo-3-trifluoromethylbenzoicacid
C1=CC(=C(C(=C1)C(F)(F)F)Br)C(=O)O
269.017
269.02
98%
Videos
SDS