missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-4,5-difluorophenylacetic acid, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
2-Bromo-4,5-difluorophenylacetic acid is used as intermediate in organic syntheses.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Bromo-4,5-difluorophenylacetic acid is used as intermediate in organic syntheses.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
2-Bromo-4,5-difluorophenylacetic acid is used as intermediate in organic syntheses.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Color | White |
Quantity | 250 mg |
Solubility Information | Slightly soluble in water. |
Formula Weight | 251.03 |
Percent Purity | 98% |
Physical Form | Solid |
Chemical Name or Material | 2-Bromo-4,5-difluorophenylacetic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'