Läs mer
2-Bromo-4-methylpentanoic Acid Methyl Ester, TRC
Produktkod. 30348067
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30348067 500 mg
1 options
Produktkod. 30348067 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB685670500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Bromo-4-methylpentanoic Acid Methyl Ester
CAS 61837-46-5
Molekylformel C7 H13 Br O2
Kvantitet 500 mg
Synonym 2-Bromo-4-methylpentanoic Acid Methyl Ester
InChI formel InChI=1S/C7H13BrO2/c1-5(2)4-6(8)7(9)10-3/h5-6H,4H2,1-3H3
LEDER COC(=O)C(Br)CC(C)C
IUPAC-namn methyl 2-bromo-4-methylpentanoate
Molekylvikt (g/mol) 209.081
Formel vikt 208.01