missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-methylpyridine, 99%, Thermo Scientific Chemicals

Catalog Number p-75484
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
4926-28-7
C6H6BrN
172.03
MFCD00082590
LSZMVESSGLHDJE-UHFFFAOYSA-N
2-bromo-4-picoline, 2-bromo-4-methyl-pyridine, 4-methyl-2-bromopyridine, pyridine, 2-bromo-4-methyl, 2 bromo-4-methyl pyridine, 2-bromo-4-methyl pyridine, pubchem1184, acmc-209kfi, 2-bromo4-methylpyridine
2734087
2-bromo-4-methylpyridine
CC1=CC=NC(Br)=C1
This item is not returnable. View return policy
4926-28-7
C6H6BrN
172.03
MFCD00082590
LSZMVESSGLHDJE-UHFFFAOYSA-N
2-bromo-4-picoline, 2-bromo-4-methyl-pyridine, 4-methyl-2-bromopyridine, pyridine, 2-bromo-4-methyl, 2 bromo-4-methyl pyridine, 2-bromo-4-methyl pyridine, pubchem1184, acmc-209kfi, 2-bromo4-methylpyridine
2734087
2-bromo-4-methylpyridine
CC1=CC=NC(Br)=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Color Yellow
Density 1.5450g/mL
Boiling Point 87°C (10.0 mmHg)
Flash Point >109°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5600 to 1.563
Quantity 1 g
Specific Gravity 1.545
Formula Weight 172.02
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Bromo-4-methylpyridine, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromo-4-methylpyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'