missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-4-pyridinecarboxaldehyde, 98%, Thermo Scientific™
Click to view available options
Quantity:
1 g
Specifications
CAS | 118289-17-1 |
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Melting Point | 54.0°C to 55.0°C |
Color | Yellow |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (GC) |
Packaging | Glass Bottle |
Molecular Formula | C6H4BrNO |
MDL Number | MFCD04039311 |
Show More |
Safety and Handling
Product Identifier
- 2-Bromo-4-pyridinecarboxaldehyde
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'