missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-chlorothiophene, 97%, Thermo Scientific Chemicals

Catalog Number p-7023286
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
2873-18-9
C4H2BrClS
197.47
MFCD00014523
ZFAJPWYXLYGUJU-UHFFFAOYSA-N
2-chloro-5-bromothiophene, 5-bromo-2-chlorothiophene, 2-chloro-5-bromo thiophene, thiophene, 2-bromo-5-chloro, 2-brom-5-chlorthiophen, pubchem5279, acmc-1cs3j, 2-bromo-5-chloro-thiophene, 5-bromo-2-chloro-thiophene, ksc207m3t
76133
2-bromo-5-chlorothiophene
ClC1=CC=C(Br)S1
This item is not returnable. View return policy
2873-18-9
C4H2BrClS
197.47
MFCD00014523
ZFAJPWYXLYGUJU-UHFFFAOYSA-N
2-chloro-5-bromothiophene, 5-bromo-2-chlorothiophene, 2-chloro-5-bromo thiophene, thiophene, 2-bromo-5-chloro, 2-brom-5-chlorthiophen, pubchem5279, acmc-1cs3j, 2-bromo-5-chloro-thiophene, 5-bromo-2-chloro-thiophene, ksc207m3t
76133
2-bromo-5-chlorothiophene
ClC1=CC=C(Br)S1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.803
Boiling Point 69°C to 70°C (18 mmHg)
Flash Point 85°C (185°F)
Refractive Index 1.596
Quantity 5 g
UN Number UN2810
Beilstein 109897
Formula Weight 197.48
Percent Purity 97%
Chemical Name or Material 2-Bromo-5-chlorothiophene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'