Learn More
2-Bromo-5-fluorobenzaldehyde, 98%, Thermo Scientific Chemicals
Description
2-Bromo-5-fluorobenzaldehyde is used as a precursor for the synthesis of 5-fluoro-3-substituted benzoxaboroles, which is used in material science as molecular receptors, building block in crystal engineering, as steroid conjugates for molecular imprinting , dyes and biosensors of alpha hydroxyl carboxylic acids. It is also used in the synthesis of 5-arylindazolo[3,2-b]quinazolin-7(5H)-one by reacting with 2-amino-N'-arylbenzohydrazide in the presence of Copper(I) bromide by the Ullmann-type reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Bromo-5-fluorobenzaldehyde is used as a precursor for the synthesis of 5-fluoro-3-substituted benzoxaboroles, which is used in material science as molecular receptors, building block in crystal engineering, as steroid conjugates for molecular imprinting , dyes and biosensors of alpha hydroxyl carboxylic acids. It is also used in the synthesis of 5-arylindazolo[3,2-b]quinazolin-7(5H)-one by reacting with 2-amino-N′-arylbenzohydrazide in the presence of Copper(I) bromide by the Ullmann-type reaction.
Solubility
Soluble in methanol.
Notes
Air sensitive. Keep container tightly closed in a cool, dry and well-ventilated place.
Specifications
Melting Point | 51°C to 56°C |
Flash Point | 87°C (190°F) |
Quantity | 5 g |
Sensitivity | Air Sensitive |
Solubility Information | Soluble in methanol. |
Formula Weight | 203.01 |
Percent Purity | 98% |
Chemical Name or Material | 2-Bromo-5-fluorobenzaldehyde |
RUO – Research Use Only