missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-fluorobenzoic acid, 99%, Thermo Scientific™

Catalog Number p-7077283
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
394-28-5
C7H3BrFO2
218.00
MFCD00142874
OQBMJMJZMDBQSM-UHFFFAOYSA-M
benzoic acid, 2-bromo-5-fluoro, 5-fluoro-2-bromobenzoic acid, 2-bromo-5-fluoro-benzoic acid, 2-bromo-5-fluorobenzoicacid, 2-bromo-5-fluorbenzoic acid, buttpark 19\01-66, pubchem1315, acmc-1ahhq, ksc226g3p, rarechem al bo 0747
2778181
2-bromo-5-fluorobenzoic acid
[O-]C(=O)C1=CC(F)=CC=C1Br
This item is not returnable. View return policy
394-28-5
C7H3BrFO2
218.00
MFCD00142874
OQBMJMJZMDBQSM-UHFFFAOYSA-M
benzoic acid, 2-bromo-5-fluoro, 5-fluoro-2-bromobenzoic acid, 2-bromo-5-fluoro-benzoic acid, 2-bromo-5-fluorobenzoicacid, 2-bromo-5-fluorbenzoic acid, buttpark 19\01-66, pubchem1315, acmc-1ahhq, ksc226g3p, rarechem al bo 0747
2778181
2-bromo-5-fluorobenzoic acid
[O-]C(=O)C1=CC(F)=CC=C1Br
Melting Point 156.0°C to 158.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 219.01
Percent Purity 99%
Chemical Name or Material 2-Bromo-5-fluorobenzoic acid
compliance-icons
Product Identifier
  • 2-Bromo-5-fluorobenzoic acid, 99%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'