Learn More
2-Bromo-5-fluorobenzonitrile, 98+%, Thermo Scientific Chemicals
Catalog Number 11471567
Click to view available options
Quantity:
5 g
25 g
57381-39-2
C7H3BrFN
200.01
MFCD00142875
MDHNVHCZDCSTMS-UHFFFAOYSA-N
2-bromo-5-fluorobenzenecarbonitrile, 2-bromo-5-fluoro-benzonitrile, benzonitrile, 2-bromo-5-fluoro, 2-cyano-4-fluorobromobenzene, pubchem3218, bromofluorobenzonitrile2, acmc-1awgr, brc6h3 f cn, ksc496i7h, 2-bromo-5-fluorobenzonitrile
93654
2-bromo-5-fluorobenzonitrile
FC1=CC=C(Br)C(=C1)C#N
57381-39-2
C7H3BrFN
200.01
MFCD00142875
MDHNVHCZDCSTMS-UHFFFAOYSA-N
2-bromo-5-fluorobenzenecarbonitrile, 2-bromo-5-fluoro-benzonitrile, benzonitrile, 2-bromo-5-fluoro, 2-cyano-4-fluorobromobenzene, pubchem3218, bromofluorobenzonitrile2, acmc-1awgr, brc6h3 f cn, ksc496i7h, 2-bromo-5-fluorobenzonitrile
93654
2-bromo-5-fluorobenzonitrile
FC1=CC=C(Br)C(=C1)C#N
2-Bromo-5-fluorobenzonitrile is used in the synthesis of pyrimidine functional compounds acting as GABAAα2/α3 binding site agonists used in the treatment of anxiety disorders.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Bromo-5-fluorobenzonitrile is used in the synthesis of pyrimidine functional compounds acting as GABAAα2/α3 binding site agonists used in the treatment of anxiety disorders.
Solubility
Soluble in methanol.
Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY
Melting Point 88°C to 92°C
Quantity 5 g
UN Number UN3439
Beilstein 8678330
Solubility Information Soluble in methanol.
Formula Weight 200.01
Percent Purity ≥98%
Chemical Name or Material 2-Bromo-5-fluorobenzonitrile