Läs mer
2-Bromo-5-fluoropyridine, 98%
Produktkod. 11368916
Change view
Kvantitet:
1 g
5 g
Förpackningsstorlek:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
11368916 5 g 5g
11358916 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 11368916 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer L19536.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
CAS: 41404-58-4 | C5H3BrFN | 175.988 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 41404-58-4
Molekylformel C5H3BrFN
Molekylvikt (g/mol) 175.988
MDL-nummer MFCD00234011
InChI-nyckel UODINHBLNPPDPD-UHFFFAOYSA-N
Synonym 2-bromo-5-fluoro-pyridine, pyridine, 2-bromo-5-fluoro, pubchem1099, pubchem14324, 2-bromo5-fluoropyridine, acmc-1assy, 2-bromo-5-fluoropyridin, 2-bromo-5-fluorpyridine, 2-bromo-5-fluropyridine, 5-fluoro-2-bromopyridine
PubChem CID 2783171
IUPAC-namn 2-bromo-5-fluoropyridine
LEDER C1=CC(=NC=C1F)Br
Specifikationer
Kemiskt namn eller material 2-Bromo-5-fluoropyridine
Smältpunkt 26°C to 31°C
Kokpunkt 80°C to 83°C (44 mmHg)
Flampunkt 73°C (163°F)
Brytningsindex 1.5396
Kvantitet 5 g
UN-nummer UN2811
Beilstein 1561456
Formel vikt 175.99
Procent renhet 98%