missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-5-hydroxybenzaldehyde, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
2-Bromo-5-hydroxybenzaldehyde is a useful reactant used in the synthesis of PDE4 inhibitors, BCL-XL and cancel cell growth inhibitors. Also an reactant used in the preparation of many pharmaceutical agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Bromo-5-hydroxybenzaldehyde is a useful reactant used in the synthesis of PDE4 inhibitors, BCL-XL and cancel cell growth inhibitors. Also an reactant used in the preparation of many pharmaceutical agents.
Solubility
Soluble in chloroform, dichloromethane and ethyl acetate.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
2-Bromo-5-hydroxybenzaldehyde is a useful reactant used in the synthesis of PDE4 inhibitors, BCL-XL and cancel cell growth inhibitors. Also an reactant used in the preparation of many pharmaceutical agents.
Solubility
Soluble in chloroform, dichloromethane and ethyl acetate.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
Melting Point | 130°C to 135°C |
Quantity | 25 g |
UN Number | UN3077 |
Beilstein | 2079921 |
Synonym | 4-bromo-3-formylphenol, 2-bromo-5-hydroxy-benzaldehyde, 2-bromo-5-hydroxy benzaldehyde, benzaldehyde, 2-bromo-5-hydroxy, 2-bromo-5-hydroxybenzadehyde, benzaldehyde,2-bromo-5-hydroxy, pubchem14430, acmc-209haw, 6-brom-3-oxy-benzaldehyd |
Solubility Information | Soluble in chloroform,dichloromethane and ethyl acetate. |
Formula Weight | 201.02 |
Percent Purity | 95% |
Physical Form | Solid |
Chemical Name or Material | 2-Bromo-5-hydroxybenzaldehyde |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'