missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-methyl-1-benzothiophene, 97%, Thermo Scientific™

Catalog Number p-3075399
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
111860-00-5
C9H7BrS
227.119
MFCD05865140
HZPBPLLBZYSGBI-UHFFFAOYSA-N
2-bromo-5-methylbenzo b thiophene, benzo b thiophene,2-bromo-5-methyl, acmc-20mexj, 2-bromanyl-5-methyl-1-benzothiophene, 2-bromo-5-methyl-1-benzo b thiophene, benzo b thiophene, 2-bromo-5-methyl
2794769
2-bromo-5-methyl-1-benzothiophene
CC1=CC2=C(C=C1)SC(=C2)Br
This item is not returnable. View return policy
111860-00-5
C9H7BrS
227.119
MFCD05865140
HZPBPLLBZYSGBI-UHFFFAOYSA-N
2-bromo-5-methylbenzo b thiophene, benzo b thiophene,2-bromo-5-methyl, acmc-20mexj, 2-bromanyl-5-methyl-1-benzothiophene, 2-bromo-5-methyl-1-benzo b thiophene, benzo b thiophene, 2-bromo-5-methyl
2794769
2-bromo-5-methyl-1-benzothiophene
CC1=CC2=C(C=C1)SC(=C2)Br
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 227.12
Percent Purity 97%
Chemical Name or Material 2-Bromo-5-methyl-1-benzothiophene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'