Läs mer
2-Bromo-5-nitropyridine, 98+%, Thermo Scientific™
Produktkod. 11322737
Change view
Kvantitet:
1 g
5 g
Förpackningsstorlek:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
11322737 5 g 5g
11312737 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 11322737 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer L15661.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
2-Bromo-5-nitropyridine was used in preparation of boc-protected (piperazin-1-ylmethyl)biaryls via microwave-mediated Suzuki-Miyaura coupling with (boc-piperazin-1-ylmethyl)phenylboronic acid pinacol esters. It was also used in the synthesis of 2-pyridyl analogs. Substituted 5-nitro-2-ethynylpyridines were synthesized by the Sonogashira reaction of 2-bromo-5-5-nitropyridine with terminal acetylenes.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 4487-59-6
Molekylformel C5H3BrN2O2
Molekylvikt (g/mol) 203.00
MDL-nummer MFCD00006222
InChI-nyckel HUUFTVUBFFESEN-UHFFFAOYSA-N
Synonym 2-bromo-5-nitro pyridine, 2-bromo-5-nitro-pyridine, 5-nitro-2-bromopyridine, 2-brom-5-nitropyridin, pubchem1183, acmc-1agnm, 2-bromo-5-nitro-pyridin, ksc237s9f, tpc-py095
PubChem CID 78240
IUPAC-namn 2-bromo-5-nitropyridine
LEDER [O-][N+](=O)C1=CC=C(Br)N=C1
Specifikationer
Kemiskt namn eller material 2-Bromo-5-nitropyridine
Smältpunkt 136°C to 140°C
Kokpunkt 145°C to 147°C (10 mmHg)
Kvantitet 5 g
Beilstein 120901
Löslighetsinformation Insoluble in water.
Formel vikt 203
Procent renhet ≥98%