Learn More
2-Bromo-5-(trifluoromethyl)aniline, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 454-79-5 |
| Molecular Formula | C7H5BrF3N |
| MDL Number | MFCD00007637 |
| Quantity | 5 g |
| SMILES | NC1=CC(C(F)(F)F)=CC=C1Br |
| Molecular Weight (g/mol) | 240.02 |
| Percent Purity | 98% |
| Chemical Name or Material | 2-Bromo-5-(trifluoromethyl)aniline |