Läs mer
2-Bromo-5-(trifluoromethyl)benzyl bromide, 97%, Thermo Scientific™
Produktkod. 11326579
Change view
Kvantitet:
1 g
5 g
Förpackningsstorlek:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
11326579 1 g 1g
11336579 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 11326579 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer H26182.03
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
2-Bromo-5-(trifluoromethyl)benzyl bromideis a reagent used in pharmaceutical research.
Notes
Lachrymatory. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 875664-32-7
Molekylformel C8H5Br2F3
Molekylvikt (g/mol) 317.931
MDL-nummer MFCD04973762
InChI-nyckel TXJHPBWTPBYLFP-UHFFFAOYSA-N
Synonym 1-bromo-2-bromomethyl-4-trifluoromethyl benzene, 2-bromo-5-trifluoromethyl benzyl bromide, 2-bromo-5-trifluoromethylbenzyl bromide, 4-bromo-3-bromomethyl benzotrifluoride, benzene,1-bromo-2-bromomethyl-4-1,1-dimethylethyl, 2-bromo-5-trifluoromethyl benzylbromide, 2-bromo-5-trifluoromethyl-benzylbromide, 1-bromanyl-2-bromomethyl-4-trifluoromethyl benzene
PubChem CID 2756249
IUPAC-namn 1-bromo-2-(bromomethyl)-4-(trifluoromethyl)benzene
LEDER C1=CC(=C(C=C1C(F)(F)F)CBr)Br
Specifikationer
Kemiskt namn eller material 2-Bromo-5-(trifluoromethyl)benzyl bromide
Brytningsindex 1.534
Kvantitet 1 g
UN-nummer UN3265
Formel vikt 317.93
Procent renhet 97%