Läs mer
2-Bromo-6-chloro-4-fluorophenol, 97%, Thermo Scientific™
Produktkod. p-7029287
Change view
Kvantitet:
1 g
5 g
25 g
Förpackningsstorlek:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet Förpackningsstorlek
11342719 1 g 1g
11352719 5 g 5g
11362719 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 11342719 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer H26734.03
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Detta Thermo Scientific varumärkesprodukt var ursprungligen en del av Alfa Aesar produktportfölj. Viss dokumentation och etikettinformation kan referera till det äldre varumärket. Originalet Alfa Aesar produkt-/artikelkod eller SKU-referens har inte ändrats som en del av varumärkesövergången till Thermo Scientific .
Applications
2-Bromo-6-chloro-4-fluorophenol are employed in the preparation of resins, dyes, explosives, lubricants, and plastics. Also they are are endowed with antioxidant property and are used as antioxidants in many formulations such as pharmaceuticals, cosmetics, electrical transformer oil, solvents, and organic reactions.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 886499-83-8
Molekylformel C6H3BrClFO
Molekylvikt (g/mol) 225.44
MDL-nummer MFCD04115954
InChI-nyckel CRWILTIFGXJONJ-UHFFFAOYSA-N
PubChem CID 3818635
IUPAC-namn 2-bromo-6-chloro-4-fluorophenol
LEDER OC1=C(Cl)C=C(F)C=C1Br
Specifikationer
Kemiskt namn eller material 2-Bromo-6-chloro-4-fluorophenol
Smältpunkt 38°C to 40°C
Kvantitet 1 g
Formel vikt 225.45
Procent renhet 97%