Läs mer
(2-Bromo-6-fluorophenyl)carbamimidothioic Acid Methyl Ester Monohydriodide, TRC
Produktkod. 30343762
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30343762 250 mg
30347511 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30343762 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB685220250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (2-Bromo-6-fluorophenyl)carbamimidothioic Acid Methyl Ester Monohydriodide
CAS 65896-13-1
Molekylformel C8H9BrFIN2S
Kvantitet 250 mg
InChI formel InChI=1S/C8H8BrFN2S.HI/c1-13-8(11)12-7-5(9)3-2-4-6(7)10;/h2-4H,1H3,(H2,11,12);1H
LEDER I.CSC(=N)Nc1c(F)cccc1Br
IUPAC-namn methyl N-(2-bromo-6-fluorophenyl)carbamimidothioate;hydroiodide
Molekylvikt (g/mol) 391.04
Formel vikt 389.8699